Learn More
2,6-Difluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2280503
Description
2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
2,6-Difluorobenzeneboronic acid | |
∼140°C (decomposition) | |
MFCD00792436 | |
8545931 | |
Soluble in methanol. | |
B(C1=C(C=CC=C1F)F)(O)O | |
157.911 | |
157.91 |
162101-25-9 | |
C6H5BF2O2 | |
1 g | |
2,6-difluorobenzeneboronic acid, 2,6-difluorophenyl boronic acid, 2,6-difluorophenyl boranediol, boronic acid, 2,6-difluorophenyl, 2,6-difluorphenylboronic acid, 2,6-difluoro phenylboric acid, boronic acid, b-2,6-difluorophenyl, 2,6-difluorophenylboricacid, pubchem1816, acmc-1celt | |
DBZAICSEFBVFHL-UHFFFAOYSA-N | |
(2,6-difluorophenyl)boronic acid | |
2734336 | |
98% |
Chemical Identifiers
162101-25-9 | |
157.911 | |
DBZAICSEFBVFHL-UHFFFAOYSA-N | |
2734336 | |
B(C1=C(C=CC=C1F)F)(O)O |
C6H5BF2O2 | |
MFCD00792436 | |
2,6-difluorobenzeneboronic acid, 2,6-difluorophenyl boronic acid, 2,6-difluorophenyl boranediol, boronic acid, 2,6-difluorophenyl, 2,6-difluorphenylboronic acid, 2,6-difluoro phenylboric acid, boronic acid, b-2,6-difluorophenyl, 2,6-difluorophenylboricacid, pubchem1816, acmc-1celt | |
(2,6-difluorophenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only