missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Difluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2280506

 View more versions of this product

Catalog No. AAB2280506


Explore available promotions

Description

Description

2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2,6-Difluorobenzeneboronic acid
∼140°C (decomposition)
MFCD00792436
8545931
Soluble in methanol.
B(C1=C(C=CC=C1F)F)(O)O
157.911
157.91
162101-25-9
C6H5BF2O2
5 g
2,6-difluorobenzeneboronic acid, 2,6-difluorophenyl boronic acid, 2,6-difluorophenyl boranediol, boronic acid, 2,6-difluorophenyl, 2,6-difluorphenylboronic acid, 2,6-difluoro phenylboric acid, boronic acid, b-2,6-difluorophenyl, 2,6-difluorophenylboricacid, pubchem1816, acmc-1celt
DBZAICSEFBVFHL-UHFFFAOYSA-N
(2,6-difluorophenyl)boronic acid
2734336
98%

Chemical Identifiers

162101-25-9
157.911
DBZAICSEFBVFHL-UHFFFAOYSA-N
2734336
B(C1=C(C=CC=C1F)F)(O)O
C6H5BF2O2
MFCD00792436
2,6-difluorobenzeneboronic acid, 2,6-difluorophenyl boronic acid, 2,6-difluorophenyl boranediol, boronic acid, 2,6-difluorophenyl, 2,6-difluorphenylboronic acid, 2,6-difluoro phenylboric acid, boronic acid, b-2,6-difluorophenyl, 2,6-difluorophenylboricacid, pubchem1816, acmc-1celt
(2,6-difluorophenyl)boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only