missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluorobenzylamine 98.0+%, TCI America™
$111.84 - $417.56
Chemical Identifiers
| CAS | 69385-30-4 |
|---|---|
| Molecular Formula | C7H8F2N |
| Molecular Weight (g/mol) | 144.14 |
| MDL Number | MFCD00010144 |
| InChI Key | PQCUDKMMPTXMAL-UHFFFAOYSA-O |
| PubChem CID | 123563 |
| IUPAC Name | (2,6-difluorophenyl)methanaminium |
| SMILES | [NH3+]CC1=C(F)C=CC=C1F |
Chemical Identifiers
| 69385-30-4 | |
| 144.14 | |
| PQCUDKMMPTXMAL-UHFFFAOYSA-O | |
| (2,6-difluorophenyl)methanaminium |
| C7H8F2N | |
| MFCD00010144 | |
| 123563 | |
| [NH3+]CC1=C(F)C=CC=C1F |
Specifications
| 69385-30-4 | |
| 177°C | |
| MFCD00010144 | |
| 2735 | |
| PQCUDKMMPTXMAL-UHFFFAOYSA-O | |
| (2,6-difluorophenyl)methanaminium | |
| 123563 | |
| ≥98.0% (GC,T) | |
| 2,6-Difluorobenzylamine |
| Yellow | |
| C7H8F2N | |
| 5 g | |
| 2,6-difluorobenzylamine, 2,6-difluorophenyl methanamine, 2,6-difluorobenzyl amine, benzenemethanamine, 2,6-difluoro, 1-2,6-difluorophenyl methanamine, 2,6-difluorophenyl methylamine, 2, 6-difluorobenzylamine, pubchem2235, 2,6-difluoro-benzylamine, acmc-1b2iy | |
| [NH3+]CC1=C(F)C=CC=C1F | |
| 144.14 | |
| 143.14 | |
| Liquid |
Safety and Handling
TSCA : No