Learn More
2,6-Diisopropylphenol, 97%, Thermo Scientific Chemicals
An anesthetic that exhibits direct activation of the GABA-A receptor
$53.44 - $526.77
Chemical Identifiers
CAS | 2078-54-8 |
---|---|
Molecular Formula | C12H18O |
Molecular Weight (g/mol) | 178.275 |
MDL Number | MFCD00008885 |
InChI Key | OLBCVFGFOZPWHH-UHFFFAOYSA-N |
Synonym | propofol, 2,6-diisopropylphenol, diprivan, disoprofol, disoprivan, diisopropylphenol, ampofol, 2,6-bis 1-methylethyl phenol, fresofol, propofolum |
PubChem CID | 4943 |
ChEBI | CHEBI:44915 |
IUPAC Name | 2,6-di(propan-2-yl)phenol |
SMILES | CC(C)C1=C(C(=CC=C1)C(C)C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0684114
|
Thermo Scientific Chemicals
L0684114 |
25 g |
Each for $53.44
|
|
|||||
AAL0684122
|
Thermo Scientific Chemicals
L0684122 |
100 g |
Each for $157.32
|
|
|||||
AAL0684136
|
Thermo Scientific Chemicals
L0684136 |
500 g |
Each for $526.77
|
|
|||||
Description
2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.
Solubility
Very slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
2078-54-8 | |
178.275 | |
OLBCVFGFOZPWHH-UHFFFAOYSA-N | |
4943 | |
2,6-di(propan-2-yl)phenol |
C12H18O | |
MFCD00008885 | |
propofol, 2,6-diisopropylphenol, diprivan, disoprofol, disoprivan, diisopropylphenol, ampofol, 2,6-bis 1-methylethyl phenol, fresofol, propofolum | |
CHEBI:44915 | |
CC(C)C1=C(C(=CC=C1)C(C)C)O |
Specifications
2078-54-8 | |
0.962g/mL | |
C12H18O | |
MFCD00008885 | |
1866484 | |
propofol, 2,6-diisopropylphenol, diprivan, disoprofol, disoprivan, diisopropylphenol, ampofol, 2,6-bis 1-methylethyl phenol, fresofol, propofolum | |
CC(C)C1=C(C(=CC=C1)C(C)C)O | |
178.275 | |
CHEBI:44915 | |
97% |
17°C to 18°C | |
254°C to 256°C | |
1.5140 | |
25 g | |
14,7834 | |
OLBCVFGFOZPWHH-UHFFFAOYSA-N | |
2,6-di(propan-2-yl)phenol | |
4943 | |
178.28 | |
2,6-Diisopropylphenol |
Safety and Handling
GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
RTECSNumber : SL0810000
TSCA : TSCA
RUO – Research Use Only