Learn More
2,6-Dimethyl-p-benzoquinone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1134214
Description
2,6-Dimethyl-p-benzoquinone is used as organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dimethyl-p-benzoquinone is used as organic intermediates.
Solubility
Soluble in chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and reducing agents.
Specifications
2,6-Dimethyl-p-benzoquinone | |
69°C to 71°C | |
MFCD00001605 | |
2041345 | |
Soluble in chloroform. Insoluble in water. | |
CC1=CC(=O)C=C(C1=O)C | |
136.15 | |
136.15 |
527-61-7 | |
C8H8O2 | |
25 g | |
2,6-dimethylbenzoquinone, 2,6-dimethyl-1,4-benzoquinone, 2,6-dimethyl-p-benzoquinone, m-xyloquinone, 2,6-xyloquinone, 2,6-dimethylquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-dimethyl, 3,5-dimethylbenzoquinone, p-benzoquinone, 2,6-dimethyl, 2,6-dimethyl-1,4-quinone | |
SENUUPBBLQWHMF-UHFFFAOYSA-N | |
2,6-dimethylcyclohexa-2,5-diene-1,4-dione | |
68241 | |
99% |
Chemical Identifiers
527-61-7 | |
136.15 | |
SENUUPBBLQWHMF-UHFFFAOYSA-N | |
68241 | |
CC1=CC(=O)C=C(C1=O)C |
C8H8O2 | |
MFCD00001605 | |
2,6-dimethylbenzoquinone, 2,6-dimethyl-1,4-benzoquinone, 2,6-dimethyl-p-benzoquinone, m-xyloquinone, 2,6-xyloquinone, 2,6-dimethylquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-dimethyl, 3,5-dimethylbenzoquinone, p-benzoquinone, 2,6-dimethyl, 2,6-dimethyl-1,4-quinone | |
2,6-dimethylcyclohexa-2,5-diene-1,4-dione |
Safety and Handling
GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
EINECSNumber : 208-420-8
RTECSNumber : DK4825000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only