missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2',6'-Dimethylacetanilide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2568722

 View more versions of this product

Catalog No. AAB2568722


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2',6'-Dimethylacetanilide
179°C to 183°C
MFCD00008675
n-2,6-dimethylphenyl acetamide, 2',6'-dimethylacetanilide, 2,6-dimethylacetanilide, 2',6'-acetoxylidide, acetamide, n-2,6-dimethylphenyl, n-acetyl-2,6-xylidine, n-acetyl-2,6-dimethylaniline, acetanilide, 2',6'-dimethyl, unii-68gzi4j9fm, 68gzi4j9fm
CC1=C(C(=CC=C1)C)NC(=O)C
163.22
163.22
2198-53-0
C10H13NO
100 g
NRPTXWYBRKRZES-UHFFFAOYSA-N
N-(2,6-dimethylphenyl)acetamide
16616
97%

Chemical Identifiers

2198-53-0
163.22
NRPTXWYBRKRZES-UHFFFAOYSA-N
16616
CC1=C(C(=CC=C1)C)NC(=O)C
C10H13NO
MFCD00008675
n-2,6-dimethylphenyl acetamide, 2',6'-dimethylacetanilide, 2,6-dimethylacetanilide, 2',6'-acetoxylidide, acetamide, n-2,6-dimethylphenyl, n-acetyl-2,6-xylidine, n-acetyl-2,6-dimethylaniline, acetanilide, 2',6'-dimethyl, unii-68gzi4j9fm, 68gzi4j9fm
N-(2,6-dimethylphenyl)acetamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 218-596-8

RTECSNumber : AN7470000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only