Learn More
2,6-Dimethylaniline, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1451222
Description
2,6-Dimethylaniline is used in pharmaceuticals, as dye intermediates and in organic synthesis. It is also used in the production of antioxidants, agricultural, pharmaceutical, rubber chemicals and other target organic molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dimethylaniline is used in pharmaceuticals, as dye intermediates and in organic synthesis. It is also used in the production of antioxidants, agricultural, pharmaceutical, rubber chemicals and other target organic molecules.
Solubility
Slightly miscible with water. Miscible with ethanol and diethyl ether.
Notes
It is sensitive to air and light. Incompatible withacids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates and halogens.
Specifications
2,6-Dimethylaniline | |
87-62-7 | |
216°C | |
Characteristic | |
C8H11N | |
UN1711 | |
14,10084 | |
Slightly miscible with water. Miscible with ethanol and diethyl ether. | |
CC1=CC=CC(C)=C1N | |
121.18 | |
CHEBI:28738 | |
99% |
11°C | |
0.981 | |
100 g | |
1.56 | |
MFCD00007747 | |
636332 | |
2,6-xylidine, 2-amino-m-xylene, o-xylidine, 2,6-dimethylbenzenamine, 2-amino-1,3-dimethylbenzene, 2,6-dimethylphenylamine, benzenamine, 2,6-dimethyl, 2,6-xylylamine, 2-amino-1,3-xylene, 1-amino-2,6-dimethylbenzene | |
UFFBMTHBGFGIHF-UHFFFAOYSA-N | |
2,6-dimethylaniline | |
6896 | |
121.18 |
Chemical Identifiers
87-62-7 | |
121.18 | |
UFFBMTHBGFGIHF-UHFFFAOYSA-N | |
6896 | |
2,6-dimethylaniline |
C8H11N | |
MFCD00007747 | |
2,6-xylidine, 2-amino-m-xylene, o-xylidine, 2,6-dimethylbenzenamine, 2-amino-1,3-dimethylbenzene, 2,6-dimethylphenylamine, benzenamine, 2,6-dimethyl, 2,6-xylylamine, 2-amino-1,3-xylene, 1-amino-2,6-dimethylbenzene | |
CHEBI:28738 | |
CC1=CC=CC(C)=C1N |
Safety and Handling
GHS H Statement
H351-H302-H312-H332-H315-H335-H227
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Combustible liquid.
P201-P202-P210-P235-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312-H315-H319-H335-H351
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLIDINES, LIQUID
EINECSNumber : 201-758-7
RTECSNumber : ZE9275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only