missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethylaniline, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1451236

 View more versions of this product

Catalog No. AAA1451236

Explore available promotions

Description

Description

2,6-Dimethylaniline is used in pharmaceuticals, as dye intermediates and in organic synthesis. It is also used in the production of antioxidants, agricultural, pharmaceutical, rubber chemicals and other target organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylaniline is used in pharmaceuticals, as dye intermediates and in organic synthesis. It is also used in the production of antioxidants, agricultural, pharmaceutical, rubber chemicals and other target organic molecules.

Solubility
Slightly miscible with water. Miscible with ethanol and diethyl ether.

Notes
It is sensitive to air and light. Incompatible withacids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates and halogens.
Specifications

Specifications

2,6-Dimethylaniline
87-62-7
216°C
Characteristic
C8H11N
UN1711
14,10084
Slightly miscible with water. Miscible with ethanol and diethyl ether.
CC1=CC=CC(C)=C1N
121.18
CHEBI:28738
99%
11°C
0.981
500 g
1.56
MFCD00007747
636332
2,6-xylidine, 2-amino-m-xylene, o-xylidine, 2,6-dimethylbenzenamine, 2-amino-1,3-dimethylbenzene, 2,6-dimethylphenylamine, benzenamine, 2,6-dimethyl, 2,6-xylylamine, 2-amino-1,3-xylene, 1-amino-2,6-dimethylbenzene
UFFBMTHBGFGIHF-UHFFFAOYSA-N
2,6-dimethylaniline
6896
121.18

Chemical Identifiers

87-62-7
121.18
UFFBMTHBGFGIHF-UHFFFAOYSA-N
6896
2,6-dimethylaniline
C8H11N
MFCD00007747
2,6-xylidine, 2-amino-m-xylene, o-xylidine, 2,6-dimethylbenzenamine, 2-amino-1,3-dimethylbenzene, 2,6-dimethylphenylamine, benzenamine, 2,6-dimethyl, 2,6-xylylamine, 2-amino-1,3-xylene, 1-amino-2,6-dimethylbenzene
CHEBI:28738
CC1=CC=CC(C)=C1N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302-H312-H332-H315-H335-H227
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Combustible liquid.

P201-P202-P210-P235-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312-H315-H319-H335-H351

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLIDINES, LIQUID

EINECSNumber : 201-758-7

RTECSNumber : ZE9275000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only