missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Dimethylbenzyl alcohol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6140906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,6-Dimethylbenzyl alcohol | |
C9H12O | |
5 g | |
JPEYJQDKTDVJSZ-UHFFFAOYSA-N | |
(2,6-dimethylphenyl)methanol | |
186748 | |
98% |
62285-58-9 | |
MFCD00128004 | |
2,6-dimethylphenyl methanol, 2,6-dimethylbenzyl alcohol, 2,6-dimethyl-benzenemethanol, 2,6-dimethylbenzylalcohol, benzenemethanol, 2,6-dimethyl, 2,6-dimethyl-phenyl-methanol, 2.3-dimethylbenzylalkohol, 2,6-dimethylbenzenemethanol, 2,6-dimethyl benzyl alcohol, benzenemethanol,2,6-dimethyl | |
CC1=C(C(=CC=C1)C)CO | |
136.194 | |
136.19 | |
Solid |
Chemical Identifiers
62285-58-9 | |
136.194 | |
JPEYJQDKTDVJSZ-UHFFFAOYSA-N | |
186748 | |
CC1=C(C(=CC=C1)C)CO |
C9H12O | |
MFCD00128004 | |
2,6-dimethylphenyl methanol, 2,6-dimethylbenzyl alcohol, 2,6-dimethyl-benzenemethanol, 2,6-dimethylbenzylalcohol, benzenemethanol, 2,6-dimethyl, 2,6-dimethyl-phenyl-methanol, 2.3-dimethylbenzylalkohol, 2,6-dimethylbenzenemethanol, 2,6-dimethyl benzyl alcohol, benzenemethanol,2,6-dimethyl | |
(2,6-dimethylphenyl)methanol |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only