missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Dimethylbenzyl Chloride 98.0+%, TCI America™
$152.63 - $462.86
Chemical Identifiers
CAS | 5402-60-8 |
---|---|
Molecular Formula | C9H11Cl |
Molecular Weight (g/mol) | 154.637 |
MDL Number | MFCD01310815 |
InChI Key | HPVRFWQMBYLJRL-UHFFFAOYSA-N |
PubChem CID | 221154 |
IUPAC Name | 2-(chloromethyl)-1,3-dimethylbenzene |
SMILES | CC1=C(C(=CC=C1)C)CCl |
Chemical Identifiers
5402-60-8 | |
154.637 | |
HPVRFWQMBYLJRL-UHFFFAOYSA-N | |
2-(chloromethyl)-1,3-dimethylbenzene |
C9H11Cl | |
MFCD01310815 | |
221154 | |
CC1=C(C(=CC=C1)C)CCl |
Specifications
5402-60-8 | |
100°C | |
MFCD01310815 | |
3261 | |
HPVRFWQMBYLJRL-UHFFFAOYSA-N | |
2-(chloromethyl)-1,3-dimethylbenzene | |
221154 | |
≥98.0% (GC) | |
2,6-Dimethylbenzyl Chloride |
White-Yellow | |
C9H11Cl | |
5 g | |
2,6-dimethylbenzyl chloride, 2-chloromethyl-1,3-dimethylbenzene, 2,6-dimethylbenzylchloride, 2,6-dimethyl benzylchloride, 1-chloromethyl-2,6-dimethylbenzene, 2-chloromethyl-1,3-dimethyl-benzene, 2-chloromethyl-m-xylene, benzene, 2-chloromethyl-1,3-dimethyl, 2, 6-dimethylbenzyl chloride | |
CC1=C(C(=CC=C1)C)CCl | |
154.637 | |
154.64 | |
Crystalline Lumps |
Safety and Handling
TSCA : No