Learn More
2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals
$152.52 - $620.44
Chemical Identifiers
CAS | 2769-71-3 |
---|---|
Molecular Formula | C9H9N |
Molecular Weight (g/mol) | 131.18 |
MDL Number | MFCD00013479 |
InChI Key | DNJLFZHMJDSJFN-UHFFFAOYSA-N |
Synonym | 2,6-dimethylphenyl isocyanide, 2,6-dimethylphenylisonitrile, 2,6-dimethylphenylisocyanide, 2,6-xyleneisonitrile, 2,6-xylyl isocyanide, benzene,2-isocyano-1,3-dimethyl, benzene, 2-isocyano-1,3-dimethyl-9ci, vic.-m-xylyl isocyanide, acmc-20ao80, 2,6-dimethyiphenyl isocyanide |
PubChem CID | 76009 |
IUPAC Name | 2-isocyano-1,3-dimethylbenzene |
SMILES | CC1=CC=CC(C)=C1[N+]#[C-] |
Description
2,6-Dimethylphenyl isocyanate is used in the preparation of homoleptic metallaamidine complex, derivatized bets-cyclodextrins. It is used as chiral stationary phase in normal-phase liquid chromatography. It is also involved in the preparation of tris( 2,6-dimethylphenylimido)methylrhenium (VII) and 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2769-71-3 | |
131.18 | |
DNJLFZHMJDSJFN-UHFFFAOYSA-N | |
76009 | |
CC1=CC=CC(C)=C1[N+]#[C-] |
C9H9N | |
MFCD00013479 | |
2,6-dimethylphenyl isocyanide, 2,6-dimethylphenylisonitrile, 2,6-dimethylphenylisocyanide, 2,6-xyleneisonitrile, 2,6-xylyl isocyanide, benzene,2-isocyano-1,3-dimethyl, benzene, 2-isocyano-1,3-dimethyl-9ci, vic.-m-xylyl isocyanide, acmc-20ao80, 2,6-dimethyiphenyl isocyanide | |
2-isocyano-1,3-dimethylbenzene |
Specifications
73°C to 74°C | |
1 g | |
Crystalline | |
MFCD00013479 | |
2,6-dimethylphenyl isocyanide, 2,6-dimethylphenylisonitrile, 2,6-dimethylphenylisocyanide, 2,6-xyleneisonitrile, 2,6-xylyl isocyanide, benzene,2-isocyano-1,3-dimethyl, benzene, 2-isocyano-1,3-dimethyl-9ci, vic.-m-xylyl isocyanide, acmc-20ao80, 2,6-dimethyiphenyl isocyanide | |
DNJLFZHMJDSJFN-UHFFFAOYSA-N | |
2-isocyano-1,3-dimethylbenzene | |
76009 | |
2,6-Dimethylphenyl isocyanide |
2769-71-3 | |
Unpleasant | |
C9H9N | |
UN2811 | |
Insoluble in water. | |
CC1=CC=CC(C)=C1[N+]#[C-] | |
131.18 | |
131.18 |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
H301+H311-H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 220-459-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only