Learn More
2,6-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H54138AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,6-Dimethylphenylmagnesium bromide | |
C8H9BrMg | |
100 mL | |
Air and moisture sensitive; May form a precipitate | |
BIGUVDUZGAJWQK-UHFFFAOYSA-M | |
magnesium;1,3-dimethylbenzene-2-ide;bromide | |
4517616 | |
0.5M in 2-MeTHF |
21450-64-6 | |
MFCD00192083 | |
UN2924 | |
2,6-dimethylphenylmagnesium bromide, grignard reagent, 2,6-dimethylphenylmagnesium bromide solution, 2,6-dimethylphenymagnesium bromide, bromo 2,6-dimethylphenyl magnesium, bromo 2,6-dimethylphenyl magnesium, 2,6-dimethyl-phenylmagnesium bromide, 2,6-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 2,6-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 2,6-dimethylphenylmagnesium bromide solution, 1.0 m in thf, 2,6-dimethylphenylmagnesium bromide solution, 1.0 m in thf | |
CC1=CC=CC(C)=C1[Mg]Br | |
209.37 | |
209.37 |
Chemical Identifiers
21450-64-6 | |
209.37 | |
BIGUVDUZGAJWQK-UHFFFAOYSA-M | |
4517616 | |
CC1=CC=CC(C)=C1[Mg]Br |
C8H9BrMg | |
MFCD00192083 | |
2,6-dimethylphenylmagnesium bromide, grignard reagent, 2,6-dimethylphenylmagnesium bromide solution, 2,6-dimethylphenymagnesium bromide, bromo 2,6-dimethylphenyl magnesium, bromo 2,6-dimethylphenyl magnesium, 2,6-dimethyl-phenylmagnesium bromide, 2,6-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 2,6-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 2,6-dimethylphenylmagnesium bromide solution, 1.0 m in thf, 2,6-dimethylphenylmagnesium bromide solution, 1.0 m in thf | |
magnesium;1,3-dimethylbenzene-2-ide;bromide |
Safety and Handling
GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only