Learn More
2,6-Dinitrotoluene, 97%, Thermo Scientific Chemicals
$46.07 - $139.34
Chemical Identifiers
CAS | 606-20-2 |
---|---|
Molecular Formula | C7H6N2O4 |
Molecular Weight (g/mol) | 182.135 |
MDL Number | MFCD00007158 |
InChI Key | XTRDKALNCIHHNI-UHFFFAOYSA-N |
Synonym | 2,6-dinitrotoluene, 2,6-dnt, benzene, 2-methyl-1,3-dinitro, 1-methyl-2,6-dinitrobenzene, 2,6-dinitromethylbenzene, toluene, 2,6-dinitro, rcra waste number u106, 2-methyl-1,3-dinitro-benzene, unii-gg7fav92mk, ccris 1006 |
PubChem CID | 11813 |
ChEBI | CHEBI:957 |
IUPAC Name | 2-methyl-1,3-dinitrobenzene |
SMILES | CC1=C(C=CC=C1[N+](=O)[O-])[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0085606
|
Thermo Scientific Chemicals
L0085606 |
5 g |
Each for $46.07
|
|
|||||
AAL0085614
|
Thermo Scientific Chemicals
L0085614 |
25 g |
Each for $139.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
606-20-2 | |
182.135 | |
XTRDKALNCIHHNI-UHFFFAOYSA-N | |
11813 | |
2-methyl-1,3-dinitrobenzene |
C7H6N2O4 | |
MFCD00007158 | |
2,6-dinitrotoluene, 2,6-dnt, benzene, 2-methyl-1,3-dinitro, 1-methyl-2,6-dinitrobenzene, 2,6-dinitromethylbenzene, toluene, 2,6-dinitro, rcra waste number u106, 2-methyl-1,3-dinitro-benzene, unii-gg7fav92mk, ccris 1006 | |
CHEBI:957 | |
CC1=C(C=CC=C1[N+](=O)[O-])[N+](=O)[O-] |
Specifications
606-20-2 | |
285°C | |
C7H6N2O4 | |
5 g | |
2052046 | |
XTRDKALNCIHHNI-UHFFFAOYSA-N | |
2-methyl-1,3-dinitrobenzene | |
11813 | |
182.14 | |
2,6-Dinitrotoluene |
58°C to 65°C | |
207°C (404°F) | |
MFCD00007158 | |
UN3454 | |
2,6-dinitrotoluene, 2,6-dnt, benzene, 2-methyl-1,3-dinitro, 1-methyl-2,6-dinitrobenzene, 2,6-dinitromethylbenzene, toluene, 2,6-dinitro, rcra waste number u106, 2-methyl-1,3-dinitro-benzene, unii-gg7fav92mk, ccris 1006 | |
CC1=C(C=CC=C1[N+](=O)[O-])[N+](=O)[O-] | |
182.135 | |
CHEBI:957 | |
97% |
Safety and Handling
GHS H Statement
H301-H311-H331-H350-H341-H361-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P501c
H301+H311+H331-H341-H350-H361f-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DINITROTOLUENES, SOLID
EINECSNumber : 210-106-0
RTECSNumber : XT1925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only