missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Pyridinedimethanol, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 167551000

Catalog No. AC167551000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 6-Pyridinedimethanol
97.5
Authentic
Glass bottle
(C5H3N)(CH2OH)2
100g
Solubility in water: soluble
C1=CC(=NC(=C1)CO)CO
139.15
139.15
1195-59-1
100.0
97.5% min. (GC)
C7H9NO2
MFCD00006351
2,6-pyridinedimethanol, pyridine-2,6-diyldimethanol, 2,6-bis hydroxymethyl pyridine, 6-hydroxymethyl pyridin-2-yl methanol, pyridine-2,6-dimethanol, 2,6-dihydroxymethylpyridine, 2,6-bis-hydroxymethyl pyridine, 2,6-bis-hydroxymethyl-pyridine, 2,6-pyridinyl dimethanol, 6-hydroxymethyl-2-pyridyl methan-1-ol
WWFMINHWJYHXHF-UHFFFAOYSA-N
[6-(hydroxymethyl)pyridin-2-yl]methanol
70957
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 214-803-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only