missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2',7'-Dichlorofluorescein, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1621214

 View more versions of this product

Catalog No. AAA1621214


Explore available promotions

Description

Description

2',7'-Dichlorofluorescein is used as an indicator and as a cellular antioxidant activity. It is also used for the determination of carbohydrates and hydrogen peroxide by fluorescence densitometry and chemiluminescence respectively. It is used as an indicator for argentometry by Fajans method. It is also involved in the cellular antioxidant activity assay. It is utilized as an additive in fluorescence and laser studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,7′-Dichlorofluorescein is used as an indicator and as a cellular antioxidant activity. It is also used for the determination of carbohydrates and hydrogen peroxide by fluorescence densitometry and chemiluminescence respectively. It is used as an indicator for argentometry by Fajans method. It is also involved in the cellular antioxidant activity assay. It is utilized as an additive in fluorescence and laser studies.

Solubility
Soluble in ethanol and methanol. Slightly dimethyl sulfoxide. Insoluble in water.

Notes
As a fluorescent indicatorIncompatible with strong oxidizing agents.
Specifications

Specifications

2',7'-Dichlorofluorescein
>300°C (decomposition)
MFCD00005047
2',7'-dichlorofluorescein, dichlorofluorescein, unii-56nqm5uzt1, 2,7-dichlorofluorescein, 56nqm5uzt1, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 2',7'-dichloro-3',6'-dihydroxy, 2',7'-dichloro-3',6'-dihydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, 2',7'-dichloro-3',6'-dihydroxyspiro 2-benzofuran-3,9'-xanthene-1-one, 11,16-dichloro-12,15-dihydroxyspiro 3-hydroisobenzofuran-3,9'-xanthene-1-one
VFNKZQNIXUFLBC-UHFFFAOYSA-N
2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
64944
401.2
76-54-0
C20H10Cl2O5
25 g
Soluble in ethanol and methanol. Slightly dimethyl sulfoxide. Insoluble in water.
C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl
401.195
CHEBI:51596

Chemical Identifiers

76-54-0
401.195
VFNKZQNIXUFLBC-UHFFFAOYSA-N
64944
2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
C20H10Cl2O5
MFCD00005047
2',7'-dichlorofluorescein, dichlorofluorescein, unii-56nqm5uzt1, 2,7-dichlorofluorescein, 56nqm5uzt1, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 2',7'-dichloro-3',6'-dihydroxy, 2',7'-dichloro-3',6'-dihydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, 2',7'-dichloro-3',6'-dihydroxyspiro 2-benzofuran-3,9'-xanthene-1-one, 11,16-dichloro-12,15-dihydroxyspiro 3-hydroisobenzofuran-3,9'-xanthene-1-one
CHEBI:51596
C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 200-968-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only