missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,7-Dinitro-9-fluorenone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5517614

 View more versions of this product

Catalog No. AAH5517614


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,7-Dinitro-9-fluorenone
292°C to 295°C
C13H6N2O5
25 g
HDVGAFBXTXDYIB-UHFFFAOYSA-N
2,7-dinitrofluoren-9-one
35842
97%
31551-45-8
Yellow to Orange
MFCD00001153
2,7-dinitro-9-fluorenone, 2,7-dinitrofluorenone, 9h-fluoren-9-one, 2,7-dinitro, 2,7-dinitro-9h-fluoren-9-one, fluoren-9-one, 2,7-dinitro, 9-fluorenone, 2,7-dinitro, ccris 2911, acmc-209hnh, maybridge1_004322, fluoren-9-one,7-dinitro
[O-][N+](=O)C1=CC=C2C3=CC=C(C=C3C(=O)C2=C1)[N+]([O-])=O
270.20
270.2
Powder

Chemical Identifiers

31551-45-8
270.20
HDVGAFBXTXDYIB-UHFFFAOYSA-N
35842
[O-][N+](=O)C1=CC=C2C3=CC=C(C=C3C(=O)C2=C1)[N+]([O-])=O
C13H6N2O5
MFCD00001153
2,7-dinitro-9-fluorenone, 2,7-dinitrofluorenone, 9h-fluoren-9-one, 2,7-dinitro, 2,7-dinitro-9h-fluoren-9-one, fluoren-9-one, 2,7-dinitro, 9-fluorenone, 2,7-dinitro, ccris 2911, acmc-209hnh, maybridge1_004322, fluoren-9-one,7-dinitro
2,7-dinitrofluoren-9-one
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 250-695-1

RTECSNumber : LL9051000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only