2,8-Bis(9H-carbazol-9-yl)dibenzothiophene 98.0+%, TCI America™
Supplier:  TCI America B49421G
 View more versions of this product
Catalog No. B49421G
Explore available promotions
Description
Description
Specifications
Specifications
2,8-Bis(9H-carbazol-9-yl)dibenzothiophene
301°C
C36H22N2S
1 g
SDHNJSIZTIODFW-UHFFFAOYSA-N
9-(8-carbazol-9-yldibenzothiophen-2-yl)carbazole
59348313
≥98.0% (HPLC,N)
913738-04-2
White-Yellow
MFCD28291905
DCzDBT, 2,8-Di(9-carbazolyl)dibenzothiophene
C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC5=C(C=C4)SC6=C5C=C(C=C6)N7C8=CC=CC=C8C9=CC=CC=C97
514.646
514.65
Crystalline Powder
Chemical Identifiers
913738-04-2
514.646
SDHNJSIZTIODFW-UHFFFAOYSA-N
59348313
C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC5=C(C=C4)SC6=C5C=C(C=C6)N7C8=CC=CC=C8C9=CC=CC=C97
C36H22N2S
MFCD28291905
DCzDBT, 2,8-Di(9-carbazolyl)dibenzothiophene
9-(8-carbazol-9-yldibenzothiophen-2-yl)carbazole
Videos
Safety and Handling
Safety and Handling
TSCA : No