missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,8-Bis(trifluoromethyl)-4-quinolinol, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 306010010

 View more versions of this product

Catalog No. AC306010010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2, 8-Bis(trifluoromethyl)-4-quinolinol
35853-41-9
100.0
98.5% min. (HPLC)
C11H5F6NO
1g
JIWHKBAFGFPZKM-UHFFFAOYSA-N
2,8-bis(trifluoromethyl)-1H-quinolin-4-one
736140
99%
99%
98.5
Authentic
Glass bottle
MFCD00075091
2,8-bis trifluoromethyl-4-hydroxyquinoline, 2,8-bis trifluoromethyl quinolin-4-ol, 2,8-bis trifluoromethyl-4-quinolinol, 2,8-bis trifluoromethyl-1h-quinolin-4-one, 2,8-bis-trifluoromethyl-quinolin-4-ol, cc-pmlsc-dma-p104, 4-quinolinol, 2,8-bis trifluoromethyl, 2,8-bis trifluoromethyl-4-hydroxy quinoleine, 2,8-di trifluoromethyl quinolin-4-ol, pubchem5934
C1=CC2=C(C(=C1)C(F)(F)F)NC(=CC2=O)C(F)(F)F
281.15
281.15

Chemical Identifiers

35853-41-9
281.15
JIWHKBAFGFPZKM-UHFFFAOYSA-N
736140
C1=CC2=C(C(=C1)C(F)(F)F)NC(=CC2=O)C(F)(F)F
C11H5F6NO
MFCD00075091
2,8-bis trifluoromethyl-4-hydroxyquinoline, 2,8-bis trifluoromethyl quinolin-4-ol, 2,8-bis trifluoromethyl-4-quinolinol, 2,8-bis trifluoromethyl-1h-quinolin-4-one, 2,8-bis-trifluoromethyl-quinolin-4-ol, cc-pmlsc-dma-p104, 4-quinolinol, 2,8-bis trifluoromethyl, 2,8-bis trifluoromethyl-4-hydroxy quinoleine, 2,8-di trifluoromethyl quinolin-4-ol, pubchem5934
2,8-bis(trifluoromethyl)-1H-quinolin-4-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 252-762-0

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only