missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Acetamidofluorene 98.0+%, TCI America™
$169.56 - $487.89
Chemical Identifiers
CAS | 53-96-3 |
---|---|
Molecular Formula | C15H13NO |
Molecular Weight (g/mol) | 223.28 |
MDL Number | MFCD00001116 |
InChI Key | CZIHNRWJTSTCEX-UHFFFAOYSA-N |
Synonym | 2-acetamidofluorene, 2-acetylaminofluorene, n-2-fluorenylacetamide, 2-aaf, n-9h-fluoren-2-yl acetamide, acetamide, n-9h-fluoren-2-yl, n-fluoren-2-ylacetamide, n-acetyl-2-aminofluorene, 2-acetaminofluorene, 2-acetylamino fluorene |
PubChem CID | 5897 |
ChEBI | CHEBI:17356 |
IUPAC Name | N-(9H-fluoren-2-yl)acetamide |
SMILES | CC(=O)NC1=CC=C2C(CC3=CC=CC=C23)=C1 |
Chemical Identifiers
53-96-3 | |
223.28 | |
CZIHNRWJTSTCEX-UHFFFAOYSA-N | |
5897 | |
N-(9H-fluoren-2-yl)acetamide |
C15H13NO | |
MFCD00001116 | |
2-acetamidofluorene, 2-acetylaminofluorene, n-2-fluorenylacetamide, 2-aaf, n-9h-fluoren-2-yl acetamide, acetamide, n-9h-fluoren-2-yl, n-fluoren-2-ylacetamide, n-acetyl-2-aminofluorene, 2-acetaminofluorene, 2-acetylamino fluorene | |
CHEBI:17356 | |
CC(=O)NC1=CC=C2C(CC3=CC=CC=C23)=C1 |
Specifications
53-96-3 | |
White-Yellow | |
MFCD00001116 | |
2-acetamidofluorene, 2-acetylaminofluorene, n-2-fluorenylacetamide, 2-aaf, n-9h-fluoren-2-yl acetamide, acetamide, n-9h-fluoren-2-yl, n-fluoren-2-ylacetamide, n-acetyl-2-aminofluorene, 2-acetaminofluorene, 2-acetylamino fluorene | |
CC(=O)NC1=CC=C2C(CC3=CC=CC=C23)=C1 | |
223.28 | |
CHEBI:17356 | |
≥98.0% (HPLC,N) | |
2-Acetamidofluorene |
196°C | |
C15H13NO | |
5 g | |
CZIHNRWJTSTCEX-UHFFFAOYSA-N | |
N-(9H-fluoren-2-yl)acetamide | |
5897 | |
223.28 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : AB9450000
TSCA : Yes