missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Acetonaphthone, 99%, Thermo Scientific Chemicals

$89.19 - $89.19

Chemical Identifiers

CAS 93-08-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00004108
InChI Key XSAYZAUNJMRRIR-UHFFFAOYSA-N
Synonym 2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
PubChem CID 7122
ChEBI CHEBI:52364
IUPAC Name 1-naphthalen-2-ylethanone
SMILES CC(=O)C1=CC2=CC=CC=C2C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC158881000
SDS View Documents
Thermo Scientific Chemicals
158881000
100 g
Each for $89.19
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

93-08-3
170.21
XSAYZAUNJMRRIR-UHFFFAOYSA-N
7122
1-naphthalen-2-ylethanone
C12H10O
MFCD00004108
2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
CHEBI:52364
CC(=O)C1=CC2=CC=CC=C2C=C1

Specifications

93-08-3
100.0
White
168°C
98.5% min. (GC)
C12H10O
MFCD00004108
07, 402
Solubility in water: insoluble
CC(=O)C1=CC2=CC=CC=C2C=C1
170.21
CHEBI:52364
99%
2'-Acetonaphthone, 99%
98.5
53.0°C to 56.0°C
300.0°C to 301.0°C
Authentic
Plastic bottle
C10H7COCH3
100 g
2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
XSAYZAUNJMRRIR-UHFFFAOYSA-N
1-naphthalen-2-ylethanone
7122
170.21
Fine Crystalline Powder and Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Warning

EINECSNumber : 202-216-2

RTECSNumber : AL2988000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only