missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Acetoxybenzonitrile 97.0+%, TCI America™
$62.29 - $192.05
Chemical Identifiers
CAS | 5715-02-6 |
---|---|
Molecular Formula | C9H7NO2 |
Molecular Weight (g/mol) | 161.16 |
MDL Number | MFCD00017360 |
InChI Key | XXLKCUTUGWSJJO-UHFFFAOYSA-N |
PubChem CID | 79791 |
IUPAC Name | (2-cyanophenyl) acetate |
SMILES | CC(=O)OC1=CC=CC=C1C#N |
Chemical Identifiers
5715-02-6 | |
161.16 | |
XXLKCUTUGWSJJO-UHFFFAOYSA-N | |
(2-cyanophenyl) acetate |
C9H7NO2 | |
MFCD00017360 | |
79791 | |
CC(=O)OC1=CC=CC=C1C#N |
Specifications
5715-02-6 | |
254°C | |
MFCD00017360 | |
2-acetoxybenzonitrile, benzonitrile,2-acetyloxy, o-acetoxybenzonitrile, pubchem13326, 2-cyanophenyl acetate, #, acmc-1aso4, benzonitrile, 2-acetyloxy, acetic acid 2-cyanophenyl ester | |
CC(=O)OC1=CC=CC=C1C#N | |
161.16 | |
161.16 | |
Liquid |
Yellow | |
C9H7NO2 | |
5 g | |
XXLKCUTUGWSJJO-UHFFFAOYSA-N | |
(2-cyanophenyl) acetate | |
79791 | |
≥97.0% (GC) | |
2-Acetoxybenzonitrile |
Safety and Handling
TSCA : No