missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetyl-5-bromothiophene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1482618

 View more versions of this product

Catalog No. AAA1482618


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Acetyl-5-bromothiophene
93°C to 96°C
C6H5BrOS
50 g
Light sensitive
IGBZCOWXSCWSHO-UHFFFAOYSA-N
1-(5-bromothiophen-2-yl)ethanone
79335
99%
5370-25-2
103°C (4 mmHg)
MFCD00014528
113949
2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone
CC(=O)C1=CC=C(S1)Br
205.069
205.07

Chemical Identifiers

5370-25-2
205.069
IGBZCOWXSCWSHO-UHFFFAOYSA-N
79335
CC(=O)C1=CC=C(S1)Br
C6H5BrOS
MFCD00014528
2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone
1-(5-bromothiophen-2-yl)ethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 226-363-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only