missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetyl-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 134780100

 View more versions of this product

Catalog No. AC134780100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Acetyl-6-methoxynaphthalene
3900-45-6
100.0
Beige-Yellow
98%
C13H12O2
MFCD00021643
2-acetyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethanone, 1-6-methoxy-2-naphthyl ethanone, 6-methoxy-2-acetylnaphthalene, unii-61vbh3537c, 6'-methoxy-2'-acetonaphthone, 1-6-methoxy-2-naphthyl ethan-1-one, ethanone, 1-6-methoxy-2-naphthalenyl, 2-acetyl-6-methoxy naphthalene, 6-methoxy-2-acetyl naphthalene
GGWCZBGAIGGTDA-UHFFFAOYSA-N
1-(6-methoxynaphthalen-2-yl)ethanone
77506
98%
98%
97.5
108.0°C to 109.0°C
Authentic
Glass bottle
CH3OC10H6COCH3
10 g
Solubility in water: insoluble
CC(=O)C1=CC2=C(C=C1)C=C(C=C2)OC
200.24
200.24
Powder

Chemical Identifiers

3900-45-6
200.24
GGWCZBGAIGGTDA-UHFFFAOYSA-N
77506
CC(=O)C1=CC2=C(C=C1)C=C(C=C2)OC
C13H12O2
MFCD00021643
2-acetyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethanone, 1-6-methoxy-2-naphthyl ethanone, 6-methoxy-2-acetylnaphthalene, unii-61vbh3537c, 6'-methoxy-2'-acetonaphthone, 1-6-methoxy-2-naphthyl ethan-1-one, ethanone, 1-6-methoxy-2-naphthalenyl, 2-acetyl-6-methoxy naphthalene, 6-methoxy-2-acetyl naphthalene
1-(6-methoxynaphthalen-2-yl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 223-453-8

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only