missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetyl-7-hydroxybenzofuran 98.0+%, TCI America™

$150.52 - $247.40

Chemical Identifiers

CAS 40020-87-9
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
MDL Number MFCD00191373
InChI Key HRIVIHCUSKDPNK-UHFFFAOYSA-N
Synonym 2-Acetyl-7-hydroxycoumarone
PubChem CID 595329
IUPAC Name 1-(7-hydroxy-1-benzofuran-2-yl)ethan-1-one
SMILES CC(=O)C1=CC2=CC=CC(O)=C2O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A13165G
View Documents
TCI America
A13165G
5 g
Each for $150.52
 
A131610G
View Documents
TCI America
A131610G
10 g
Each for $247.40
 
Description

Description

Specifications

Chemical Identifiers

40020-87-9
176.17
HRIVIHCUSKDPNK-UHFFFAOYSA-N
595329
CC(=O)C1=CC2=CC=CC(O)=C2O1
C10H8O3
MFCD00191373
2-Acetyl-7-hydroxycoumarone
1-(7-hydroxy-1-benzofuran-2-yl)ethan-1-one

Specifications

40020-87-9
Yellow
MFCD00191373
2-Acetyl-7-hydroxycoumarone
CC(=O)C1=CC2=CC=CC(O)=C2O1
176.17
176.17
Crystalline Powder
166°C
C10H8O3
5 g
HRIVIHCUSKDPNK-UHFFFAOYSA-N
1-(7-hydroxy-1-benzofuran-2-yl)ethan-1-one
595329
≥98.0% (GC)
2-Acetyl-7-hydroxybenzofuran
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'