missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetylnaphthalene, 99%, Thermo Scientific Chemicals

$51.14 - $153.03

Chemical Identifiers

CAS 93-08-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00004108
InChI Key XSAYZAUNJMRRIR-UHFFFAOYSA-N
Synonym 2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
PubChem CID 7122
ChEBI CHEBI:52364
IUPAC Name 1-naphthalen-2-ylethanone
SMILES CC(=O)C1=CC2=CC=CC=C2C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1479318
View Documents
Thermo Scientific Chemicals
A1479318
50 g
Each for $51.14
 
AAA1479322
View Documents
Thermo Scientific Chemicals
A1479322
100 g
Each for $88.95
 
AAA1479330
View Documents
Thermo Scientific Chemicals
A1479330
250 g
Each for $153.03
 
Description

Description

2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.

Solubility
Soluble in ethanol, acetone and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications

Chemical Identifiers

93-08-3
170.211
XSAYZAUNJMRRIR-UHFFFAOYSA-N
7122
1-naphthalen-2-ylethanone
C12H10O
MFCD00004108
2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
CHEBI:52364
CC(=O)C1=CC2=CC=CC=C2C=C1

Specifications

93-08-3
300°C to 301°C
C12H10O
50 g
774965
Soluble in ethanol,acetone and ether. Insoluble in water.
CC(=O)C1=CC2=CC=CC=C2C=C1
170.211
CHEBI:52364
99%
52°C to 56°C
168°C (334°F)
MFCD00004108
UN3077
2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
XSAYZAUNJMRRIR-UHFFFAOYSA-N
1-naphthalen-2-ylethanone
7122
170.21
2-Acetylnaphthalene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 202-216-2

RTECSNumber : AL2988000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only