missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyridine, 98%

CAS: 1122-62-9 | C7H7NO | 121.14 g/mol

$87.60 - $290.04

Chemical Identifiers

CAS 1122-62-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
InChI Key AJKVQEKCUACUMD-UHFFFAOYSA-N
Synonym 2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone
PubChem CID 14286
IUPAC Name 1-pyridin-2-ylethanone
SMILES CC(=O)C1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC102680250
SDS View Documents
Thermo Scientific Chemicals
102680250
25 g Glass bottle
Each for $87.60
Only null left
 
AC102681000
SDS View Documents
Thermo Scientific Chemicals
102681000
100 g Glass bottle
Each for $290.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1122-62-9
121.14
2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone
1-pyridin-2-ylethanone
C7H7NO
AJKVQEKCUACUMD-UHFFFAOYSA-N
14286
CC(=O)C1=CC=CC=N1

Specifications

1122-62-9
100.0
Colorless to Brown
188.0°C to 189.0°C
Authentic
Glass bottle
1.5200 to 1.5220
25 g
2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone
AJKVQEKCUACUMD-UHFFFAOYSA-N
1-pyridin-2-ylethanone
14286
98%
2-Acetylpyridine
97.5
8.0°C to 10.0°C
1.0800g/mL
76°C
97.5% min. (GC)
C7H7NO
C5H4NCOCH3
1.08
Solubility in water: 200g/L (20°C). Other solubilities: miscible with ethanol
CC(=O)C1=CC=CC=N1
121.14
121.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 214-355-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only