missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-[(Acetylthio)methyl]-3-phenylpropionic Acid 98.0+%, TCI America™
$120.48 - $365.38
Chemical Identifiers
CAS | 91702-98-6 |
---|---|
Molecular Formula | C12H14O3S |
Molecular Weight (g/mol) | 238.301 |
MDL Number | MFCD00236751 |
InChI Key | BCAAXVOKLXDSPD-UHFFFAOYSA-N |
PubChem CID | 9991796 |
IUPAC Name | 2-(acetylsulfanylmethyl)-3-phenylpropanoic acid |
SMILES | CC(=O)SCC(CC1=CC=CC=C1)C(=O)O |
Chemical Identifiers
91702-98-6 | |
238.301 | |
BCAAXVOKLXDSPD-UHFFFAOYSA-N | |
2-(acetylsulfanylmethyl)-3-phenylpropanoic acid |
C12H14O3S | |
MFCD00236751 | |
9991796 | |
CC(=O)SCC(CC1=CC=CC=C1)C(=O)O |
Specifications
91702-98-6 | |
White-Yellow | |
MFCD00236751 | |
BCAAXVOKLXDSPD-UHFFFAOYSA-N | |
2-(acetylsulfanylmethyl)-3-phenylpropanoic acid | |
9991796 | |
≥98.0% (T) | |
2-[(Acetylthio)methyl]-3-phenylpropionic Acid |
39°C | |
C12H14O3S | |
1 g | |
CC(=O)SCC(CC1=CC=CC=C1)C(=O)O | |
238.301 | |
238.30 | |
Crystalline Lumps |
Safety and Handling
TSCA : No