missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Adamantylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H58595AD

Catalog No. AAH58595AD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Adamantylzinc bromide used as a intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry
Specifications

Specifications

2-Adamantylzinc bromide
Black to Yellow
ChemSeal™ bottles
MFCD01311401
UN3399
2-adamantylzinc bromide solution, adamantan-2-yl bromo zinc, adamantan-2-ide,bromozinc 1+, 2-adamantylzinc bromide,0.5m solution i, 2-adamantylzinc bromide solution, 0.5 m in thf, 2-adamantylzinc bromide 0.5 m in tetrahydrofuran, bromo 1r,3r,5r,7r-adamantan-2-yl zinc, 2-adamantylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
UKVCFEOZGIQMNI-UHFFFAOYSA-M
adamantan-2-ide;bromozinc(1+)
4145216
Liquid
171860-65-4
1.018 g/mL
C10H15BrZn
50 mL
Air sensitive; May form a precipitate
Reacts with water.
C1C2CC3CC1CC(C2)[CH-]3.[Zn+]Br
280.514
280.51

Chemical Identifiers

171860-65-4
280.514
UKVCFEOZGIQMNI-UHFFFAOYSA-M
4145216
C1C2CC3CC1CC(C2)[CH-]3.[Zn+]Br
C10H15BrZn
MFCD01311401
2-adamantylzinc bromide solution, adamantan-2-yl bromo zinc, adamantan-2-ide,bromozinc 1+, 2-adamantylzinc bromide,0.5m solution i, 2-adamantylzinc bromide solution, 0.5 m in thf, 2-adamantylzinc bromide 0.5 m in tetrahydrofuran, bromo 1r,3r,5r,7r-adamantan-2-yl zinc, 2-adamantylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
adamantan-2-ide;bromozinc(1+)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H260-H302-H314-H335-H336-H351

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only