missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Allenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™
Supplier: TCI America A20865G
Specifications
2-Allenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
Yellow | |
C9H15BO2 | |
5 g | |
CJAOMXUZZONOSD-UHFFFAOYNA-N | |
4,4,5,5-tetramethyl-2-(propa-1,2-dien-1-yl)-1,3,2-dioxaborolane | |
11679867 | |
≥95.0% (GC,T) |
865350-17-0 | |
81°C | |
MFCD09265121 | |
Allenylboronic Acid Pinacol Ester | |
CC1(C)OB(OC1(C)C)C=C=C | |
166.03 | |
166.03 | |
Liquid |
Chemical Identifiers
865350-17-0 | |
166.03 | |
CJAOMXUZZONOSD-UHFFFAOYNA-N | |
11679867 | |
CC1(C)OB(OC1(C)C)C=C=C |
C9H15BO2 | |
MFCD09265121 | |
Allenylboronic Acid Pinacol Ester | |
4,4,5,5-tetramethyl-2-(propa-1,2-dien-1-yl)-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator