missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-1-methoxybutane 90.0+%, TCI America™
$61.51 - $369.08
Chemical Identifiers
| CAS | 63448-63-5 |
|---|---|
| Molecular Formula | C5H13NO |
| Molecular Weight (g/mol) | 103.165 |
| MDL Number | MFCD00047920 |
| InChI Key | QAFODUGVXFNLBE-UHFFFAOYSA-N |
| Synonym | 1-Methoxy-sec-butylamine |
| PubChem CID | 93837 |
| IUPAC Name | 1-methoxybutan-2-amine |
| SMILES | CCC(COC)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M074625ML
|
TCI America
M074625ML |
25 mL |
Each for $61.51
|
|
|||||
|
M0746500ML
|
TCI America
M0746500ML |
500 mL |
Each for $369.08
|
|
|||||
Chemical Identifiers
| 63448-63-5 | |
| 103.165 | |
| QAFODUGVXFNLBE-UHFFFAOYSA-N | |
| 93837 | |
| CCC(COC)N |
| C5H13NO | |
| MFCD00047920 | |
| 1-Methoxy-sec-butylamine | |
| 1-methoxybutan-2-amine |
Specifications
| 63448-63-5 | |
| C5H13NO | |
| 25 mL | |
| 1-Methoxy-sec-butylamine | |
| CCC(COC)N | |
| 103.165 | |
| 103.17 | |
| Liquid |
| Yellow | |
| MFCD00047920 | |
| 2734 | |
| QAFODUGVXFNLBE-UHFFFAOYSA-N | |
| 1-methoxybutan-2-amine | |
| 93837 | |
| ≥90.0% (GC) | |
| 2-Amino-1-methoxybutane |
Safety and Handling
TSCA : Yes