Learn More
2-Amino-2-methyl-1,3-propanediol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 104051000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Amino-2-methyl-1, 3-propanediol | |
115-69-5 | |
100.0 | |
White | |
Authentic | |
Glass bottle | |
HOCH2C(CH3)(NH2)CH2OH | |
100 g | |
02,24 | |
2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol | |
UXFQFBNBSPQBJW-UHFFFAOYSA-N | |
2-amino-2-methylpropane-1,3-diol | |
1531 | |
105.14 | |
Crystalline Powder |
99% | |
98.5 | |
108.0°C to 110.0°C | |
151.0°C (10.0 mmHg) | |
99% | |
C4H11NO2 | |
MFCD00004678 | |
04, 303 | |
15, 445 | |
Solubility in water: 2500g/L (20°C). Other solubilities: soluble in alcohol | |
CC(CO)(CO)N | |
105.14 | |
CHEBI:991 | |
99% |
Chemical Identifiers
115-69-5 | |
105.14 | |
UXFQFBNBSPQBJW-UHFFFAOYSA-N | |
1531 | |
2-amino-2-methylpropane-1,3-diol |
C4H11NO2 | |
MFCD00004678 | |
2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol | |
CHEBI:991 | |
CC(CO)(CO)N |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May be corrosive to metals.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
EINECSNumber : 204-100-7
RTECSNumber : TY2975000
TSCA : TSCA
RUO – Research Use Only