missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-2-methyl-1-propanol, 95%, may cont. ca 5% water, Thermo Scientific Chemicals

$50.22 - $250.19

Chemical Identifiers

CAS 124-68-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00008051
InChI Key CBTVGIZVANVGBH-UHFFFAOYSA-N
Synonym 2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
PubChem CID 11807
IUPAC Name 2-amino-2-methylpropan-1-ol
SMILES CC(C)(N)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA17814AE
View Documents
Thermo Scientific Chemicals
A17814AE
100 mL
Each for $50.22
 
AAA17814AP
View Documents
Thermo Scientific Chemicals
A17814AP
500 mL
Each for $69.40
 
AAA178140F
View Documents
Thermo Scientific Chemicals
A178140F
2500 mL
Each for $250.19
 
Description

Description

2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-2-methyl-1-propanol is used for the preparation of buffer solution and in cosmetics. It is also used in ATR-FTIR spectroscopic investigation of the carbon monoxide absorption characteristics of a series of heterocyclic diamines.

Solubility
Immiscible with water. Miscible with alcohols.

Notes
Incompatible withoxidizing agents, strong acids, copper, brass and aluminum.
Specifications

Chemical Identifiers

124-68-5
89.14
CBTVGIZVANVGBH-UHFFFAOYSA-N
11807
CC(C)(N)CO
C4H11NO
MFCD00008051
2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
2-amino-2-methylpropan-1-ol

Specifications

124-68-5
0.934
164°C to 166°C
Amine-like
1.4455
100 mL
14,449
Immiscible with water. Miscible with alcohols.
CC(C)(N)CO
89.14
89.14
2-Amino-2-methyl-1-propanol
24°C to 28°C
11.3
67°C (152°F)
C4H11NO
MFCD00008051
505979
2-amino-2-methyl-1-propanol, isobutanol-2-amine, 1-propanol, 2-amino-2-methyl, amp regular, 2-aminoisobutanol, aminomethylpropanol, 2-amino-2-methylpropanol, 2-methyl-2-aminopropanol, 2-aminodimethylethanol, hydroxy-tert-butylamine
CBTVGIZVANVGBH-UHFFFAOYSA-N
2-amino-2-methylpropan-1-ol
11807
95%
may cont. ≈5% water
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P210-P234-P235-P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P370+P378q-P390-P501c

H227-H315-H319-H336-H373

EINECSNumber : 204-709-8

RTECSNumber : UA5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only