missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,4-dimethylbenzoic acid, 98+%

CAS: 50419-58-4 | C9H11NO2 | 165.19 g/mol

$250.67 - $916.19

Chemical Identifiers

CAS 50419-58-4
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00130041
InChI Key MUOBMUYSNYMSDM-UHFFFAOYSA-N
Synonym 2-amino-3,4-dimethyl-benzoic acid, 3,4-dimethylanthranilic acid, benzoic acid, 2-amino-3,4-dimethyl, pubchem4683, acmc-1aomr, ksc272a3h, 2-amino-3,4-dimethyl benzoic acid
PubChem CID 282450
IUPAC Name 2-amino-3,4-dimethylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(N)=C1C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAB2531306
View Documents
Thermo Scientific Chemicals
B2531306
5 g
Each for $250.67
Only null left
 
AAB2531314
View Documents
Thermo Scientific Chemicals
B2531314
25 g
Each for $916.19
Only null left
 
Description

Description

2-Amino-3,4-dimethylbenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,4-dimethylbenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications

Chemical Identifiers

50419-58-4
165.19
MUOBMUYSNYMSDM-UHFFFAOYSA-N
282450
CC1=CC=C(C(O)=O)C(N)=C1C
C9H11NO2
MFCD00130041
2-amino-3,4-dimethyl-benzoic acid, 3,4-dimethylanthranilic acid, benzoic acid, 2-amino-3,4-dimethyl, pubchem4683, acmc-1aomr, ksc272a3h, 2-amino-3,4-dimethyl benzoic acid
2-amino-3,4-dimethylbenzoic acid

Specifications

50419-58-4
C9H11NO2
5 g
Slightly soluble in water.
CC1=CC=C(C(O)=O)C(N)=C1C
165.19
165.19
2-Amino-3,4-dimethylbenzoic acid
∼176°C (decomposition)
MFCD00130041
2-amino-3,4-dimethyl-benzoic acid, 3,4-dimethylanthranilic acid, benzoic acid, 2-amino-3,4-dimethyl, pubchem4683, acmc-1aomr, ksc272a3h, 2-amino-3,4-dimethyl benzoic acid
MUOBMUYSNYMSDM-UHFFFAOYSA-N
2-amino-3,4-dimethylbenzoic acid
282450
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only