missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-3,5-dichlorobenzonitrile 98.0+%, TCI America™
$143.60 - $425.60
Chemical Identifiers
CAS | 36764-94-0 |
---|---|
Molecular Formula | C7H4Cl2N2 |
Molecular Weight (g/mol) | 187.02 |
MDL Number | MFCD00158947 |
InChI Key | ZHKNDJRPOVUPMT-UHFFFAOYSA-N |
Synonym | 2-amino-3,5-dichloro-benzonitrile, 2-amino-3,5-dichlorobenzenecarbonitrile, pubchem11850, 3,5-dichloroanthranilonitrile, buttpark 52\14-89, benzonitrile,2-amino-3,5-dichloro, 2-amino-3,5-dichlorobenzonitrile |
PubChem CID | 737215 |
IUPAC Name | 2-amino-3,5-dichlorobenzonitrile |
SMILES | NC1=C(Cl)C=C(Cl)C=C1C#N |
Chemical Identifiers
36764-94-0 | |
187.02 | |
ZHKNDJRPOVUPMT-UHFFFAOYSA-N | |
737215 | |
NC1=C(Cl)C=C(Cl)C=C1C#N |
C7H4Cl2N2 | |
MFCD00158947 | |
2-amino-3,5-dichloro-benzonitrile, 2-amino-3,5-dichlorobenzenecarbonitrile, pubchem11850, 3,5-dichloroanthranilonitrile, buttpark 52\14-89, benzonitrile,2-amino-3,5-dichloro, 2-amino-3,5-dichlorobenzonitrile | |
2-amino-3,5-dichlorobenzonitrile |
Specifications
36764-94-0 | |
White-Yellow | |
MFCD00158947 | |
2-amino-3,5-dichloro-benzonitrile, 2-amino-3,5-dichlorobenzenecarbonitrile, pubchem11850, 3,5-dichloroanthranilonitrile, buttpark 52\14-89, benzonitrile,2-amino-3,5-dichloro, 2-amino-3,5-dichlorobenzonitrile | |
NC1=C(Cl)C=C(Cl)C=C1C#N | |
187.02 | |
187.02 | |
Crystalline Powder |
124°C | |
C7H4Cl2N2 | |
1 g | |
ZHKNDJRPOVUPMT-UHFFFAOYSA-N | |
2-amino-3,5-dichlorobenzonitrile | |
737215 | |
≥98.0% (GC) | |
2-Amino-3,5-dichlorobenzonitrile |
Safety and Handling
TSCA : No