missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-3-bromobenzoic acid, 97%, Thermo Scientific Chemicals

$95.36 - $280.79

Chemical Identifiers

CAS 20776-51-6
Molecular Formula C7H5BrNO2
Molecular Weight (g/mol) 215.03
MDL Number MFCD03618453
InChI Key SRIZNTFPBWRGPB-UHFFFAOYSA-M
Synonym 3-bromoanthralic acid, 2-amino-3-bromo-benzoic acid, benzoic acid, 2-amino-3-bromo, 3-bromoanthranilic acid, 2-amino-3-bromobenzoicacid, 2-amino-3-bromo benzoic acid, buttpark 49\07-48, 2-amino-3-bromo-benzoicacid, zlchem 426
PubChem CID 270259
IUPAC Name 2-amino-3-bromobenzoic acid
SMILES NC1=C(Br)C=CC=C1C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6192503
View Documents
Thermo Scientific Chemicals
H6192503
1 g
Each for $95.36
 
AAH6192506
View Documents
Thermo Scientific Chemicals
H6192506
5 g
Each for $280.79
 
Description

Description

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

20776-51-6
215.03
SRIZNTFPBWRGPB-UHFFFAOYSA-M
270259
NC1=C(Br)C=CC=C1C([O-])=O
C7H5BrNO2
MFCD03618453
3-bromoanthralic acid, 2-amino-3-bromo-benzoic acid, benzoic acid, 2-amino-3-bromo, 3-bromoanthranilic acid, 2-amino-3-bromobenzoicacid, 2-amino-3-bromo benzoic acid, buttpark 49\07-48, 2-amino-3-bromo-benzoicacid, zlchem 426
2-amino-3-bromobenzoic acid

Specifications

20776-51-6
White to Yellow
C7H5BrNO2
1 g
3-bromoanthralic acid, 2-amino-3-bromo-benzoic acid, benzoic acid, 2-amino-3-bromo, 3-bromoanthranilic acid, 2-amino-3-bromobenzoicacid, 2-amino-3-bromo benzoic acid, buttpark 49\07-48, 2-amino-3-bromo-benzoicacid, zlchem 426
NC1=C(Br)C=CC=C1C([O-])=O
215.03
216.03
Solid
174°C to 178°C
161°C (322°F)
MFCD03618453
2936928
SRIZNTFPBWRGPB-UHFFFAOYSA-M
2-amino-3-bromobenzoic acid
270259
97%
2-Amino-3-bromobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only