Learn More
2-Amino-3-chlorobenzoic acid, 98%, Thermo Scientific Chemicals
$167.90 - $534.93
Chemical Identifiers
CAS | 6388-47-2 |
---|---|
Molecular Formula | C7H6ClNO2 |
Molecular Weight (g/mol) | 171.58 |
MDL Number | MFCD00134229 |
InChI Key | LWUAMROXVQLJKA-UHFFFAOYSA-N |
Synonym | 3-chloroanthranilic acid, benzoic acid, 2-amino-3-chloro, 2-amino-3-chloro-benzoic acid, 3-chloro-2-aminobenzoic acid, anthranilic acid, 3-chloro, 2-amino-3-chlorobenzoicacid, pubchem4639, 3-chloroanthranilicacid, acmc-209nj4, amino-3-chlorobenzoic acid |
PubChem CID | 80807 |
IUPAC Name | 2-amino-3-chlorobenzoic acid |
SMILES | NC1=C(Cl)C=CC=C1C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6626114
|
Thermo Scientific Chemicals
H6626114 |
25 g |
Each for $167.90
|
|
|||||
AAH6626122
|
Thermo Scientific Chemicals
H6626122 |
100 g |
Each for $534.93
|
|
|||||
Description
2-Amino-3-chlorobenzoic acid is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-3-chlorobenzoic acid is used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Air & Light Sensitive. Store away from air. Store in the dark. Incompatible with light, air and oxidizing agents.
Chemical Identifiers
6388-47-2 | |
171.58 | |
LWUAMROXVQLJKA-UHFFFAOYSA-N | |
80807 | |
NC1=C(Cl)C=CC=C1C(O)=O |
C7H6ClNO2 | |
MFCD00134229 | |
3-chloroanthranilic acid, benzoic acid, 2-amino-3-chloro, 2-amino-3-chloro-benzoic acid, 3-chloro-2-aminobenzoic acid, anthranilic acid, 3-chloro, 2-amino-3-chlorobenzoicacid, pubchem4639, 3-chloroanthranilicacid, acmc-209nj4, amino-3-chlorobenzoic acid | |
2-amino-3-chlorobenzoic acid |
Specifications
6388-47-2 | |
Yellow to Brown | |
MFCD00134229 | |
2090431 | |
3-chloroanthranilic acid, benzoic acid, 2-amino-3-chloro, 2-amino-3-chloro-benzoic acid, 3-chloro-2-aminobenzoic acid, anthranilic acid, 3-chloro, 2-amino-3-chlorobenzoicacid, pubchem4639, 3-chloroanthranilicacid, acmc-209nj4, amino-3-chlorobenzoic acid | |
LWUAMROXVQLJKA-UHFFFAOYSA-N | |
2-amino-3-chlorobenzoic acid | |
80807 | |
98% | |
2-Amino-3-chlorobenzoic acid |
189°C to 193°C | |
C7H6ClNO2 | |
25 g | |
Air and light sensitive | |
Soluble in water. | |
NC1=C(Cl)C=CC=C1C(O)=O | |
171.58 | |
171.58 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-996-4
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only