Learn More
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
$436.45 - $436.45
Chemical Identifiers
CAS | 852062-17-0 |
---|---|
Molecular Formula | C6H4F4N2 |
Molecular Weight (g/mol) | 180.106 |
MDL Number | MFCD10699117 |
InChI Key | HAVVQRHBCUBQGG-UHFFFAOYSA-N |
Synonym | 2-amino-3-fluoro-5-trifluoromethyl pyridine, 3-fluoro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-fluoro-5-trifluoromethylpyridine, 3-fluoro-5-trifluoromethyl-2-pyridylamine, 3-fluoro-5-trifluoromethyl pyridine-2-amine, 3-fluoro-5-trifluoromethyl-2-pyridinamine |
PubChem CID | 25067349 |
IUPAC Name | 3-fluoro-5-(trifluoromethyl)pyridin-2-amine |
SMILES | C1=C(C=NC(=C1F)N)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH3253206
|
Thermo Scientific Chemicals
H3253206 |
5 g |
Each for $436.45
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
852062-17-0 | |
180.106 | |
HAVVQRHBCUBQGG-UHFFFAOYSA-N | |
25067349 | |
C1=C(C=NC(=C1F)N)C(F)(F)F |
C6H4F4N2 | |
MFCD10699117 | |
2-amino-3-fluoro-5-trifluoromethyl pyridine, 3-fluoro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-fluoro-5-trifluoromethylpyridine, 3-fluoro-5-trifluoromethyl-2-pyridylamine, 3-fluoro-5-trifluoromethyl pyridine-2-amine, 3-fluoro-5-trifluoromethyl-2-pyridinamine | |
3-fluoro-5-(trifluoromethyl)pyridin-2-amine |
Specifications
852062-17-0 | |
C6H4F4N2 | |
5 g | |
2-amino-3-fluoro-5-trifluoromethyl pyridine, 3-fluoro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-fluoro-5-trifluoromethylpyridine, 3-fluoro-5-trifluoromethyl-2-pyridylamine, 3-fluoro-5-trifluoromethyl pyridine-2-amine, 3-fluoro-5-trifluoromethyl-2-pyridinamine | |
C1=C(C=NC(=C1F)N)C(F)(F)F | |
180.106 | |
180.1 | |
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine |
64°C to 68°C | |
MFCD10699117 | |
UN2811 | |
HAVVQRHBCUBQGG-UHFFFAOYSA-N | |
3-fluoro-5-(trifluoromethyl)pyridin-2-amine | |
25067349 | |
97% |
Safety and Handling
GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H301+H311-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only