missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-3-hydroxyanthraquinone 98.0+%, TCI America™
$128.38 - $1289.68
Chemical Identifiers
CAS | 117-77-1 |
---|---|
Molecular Formula | C14H9NO3 |
Molecular Weight (g/mol) | 239.23 |
MDL Number | MFCD00059499 |
InChI Key | CNWWMJSRHGXXAX-UHFFFAOYSA-N |
PubChem CID | 67029 |
IUPAC Name | 2-amino-3-hydroxyanthracene-9,10-dione |
SMILES | C1=CC=C2C(=C1)C(=O)C3=CC(=C(C=C3C2=O)O)N |
Chemical Identifiers
117-77-1 | |
239.23 | |
CNWWMJSRHGXXAX-UHFFFAOYSA-N | |
2-amino-3-hydroxyanthracene-9,10-dione |
C14H9NO3 | |
MFCD00059499 | |
67029 | |
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C=C3C2=O)O)N |
Specifications
117-77-1 | |
MFCD00059499 | |
CNWWMJSRHGXXAX-UHFFFAOYSA-N | |
2-amino-3-hydroxyanthracene-9,10-dione | |
67029 | |
≥98.0% (T) | |
2-Amino-3-hydroxyanthraquinone |
C14H9NO3 | |
25 g | |
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C=C3C2=O)O)N | |
239.23 | |
239.23 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0717
TSCA : No