missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-4-bromophenol 98.0+%, TCI America™
Supplier: TCI America A227125G
Specifications
2-Amino-4-bromophenol | |
133°C | |
MFCD00235171 | |
phenol, 2-amino-4-bromo, 5-bromo-2-hydroxyaniline, 4-bromo-2-aminophenol, 2-amino-4-bromo-phenol, 2-amino-4-bromophenole, 2-amino-4bromophenol, pubchem4511, acmc-209jho, 2-amino4-bromophenol, ksc493s8p | |
NC1=CC(Br)=CC=C1O | |
188.02 | |
188.02 | |
Crystalline Powder |
40925-68-6 | |
C6H6BrNO | |
25 g | |
JHRIPENGTGSNPJ-UHFFFAOYSA-N | |
2-amino-4-bromophenol | |
351840 | |
≥98.0% (GC) |
Chemical Identifiers
40925-68-6 | |
188.02 | |
JHRIPENGTGSNPJ-UHFFFAOYSA-N | |
351840 | |
NC1=CC(Br)=CC=C1O |
C6H6BrNO | |
MFCD00235171 | |
phenol, 2-amino-4-bromo, 5-bromo-2-hydroxyaniline, 4-bromo-2-aminophenol, 2-amino-4-bromo-phenol, 2-amino-4-bromophenole, 2-amino-4bromophenol, pubchem4511, acmc-209jho, 2-amino4-bromophenol, ksc493s8p | |
2-amino-4-bromophenol |
Safety and Handling
TSCA : No