missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-bromopyrimidine, 98%, Thermo Scientific Chemicals

$172.23 - $591.38

Chemical Identifiers

CAS 343926-69-2
Molecular Formula C4H4BrN3
Molecular Weight (g/mol) 174.001
MDL Number MFCD09999168
InChI Key MINURGGJJKMQQQ-UHFFFAOYSA-N
Synonym 2-amino-4-bromopyrimidine, 4-bromo-2-pyrimidinamine, 2-pyrimidinamine,4-bromo, 4-bromanylpyrimidin-2-amine, 4-bromopyrimidine-2-ylamine, 2-amino-6-bromopyrimidine, 4-bromo-2-aminopyrimidine, 2-pyrimidinamine,4-bromo-9ci, 2-pyrimidinamine, 4-bromo, 4-bromo-pyrimidin-2-ylamine
PubChem CID 12803496
IUPAC Name 4-bromopyrimidin-2-amine
SMILES C1=CN=C(N=C1Br)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64358MD
View Documents
Thermo Scientific Chemicals
H64358MD
250 mg
Each for $172.23
 
AAH6435803
View Documents
Thermo Scientific Chemicals
H6435803
1 g
Each for $591.38
 
Description

Description

It is used as a pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

343926-69-2
174.001
MINURGGJJKMQQQ-UHFFFAOYSA-N
12803496
C1=CN=C(N=C1Br)N
C4H4BrN3
MFCD09999168
2-amino-4-bromopyrimidine, 4-bromo-2-pyrimidinamine, 2-pyrimidinamine,4-bromo, 4-bromanylpyrimidin-2-amine, 4-bromopyrimidine-2-ylamine, 2-amino-6-bromopyrimidine, 4-bromo-2-aminopyrimidine, 2-pyrimidinamine,4-bromo-9ci, 2-pyrimidinamine, 4-bromo, 4-bromo-pyrimidin-2-ylamine
4-bromopyrimidin-2-amine

Specifications

343926-69-2
C4H4BrN3
250 mg
2-amino-4-bromopyrimidine, 4-bromo-2-pyrimidinamine, 2-pyrimidinamine,4-bromo, 4-bromanylpyrimidin-2-amine, 4-bromopyrimidine-2-ylamine, 2-amino-6-bromopyrimidine, 4-bromo-2-aminopyrimidine, 2-pyrimidinamine,4-bromo-9ci, 2-pyrimidinamine, 4-bromo, 4-bromo-pyrimidin-2-ylamine
MINURGGJJKMQQQ-UHFFFAOYSA-N
4-bromopyrimidin-2-amine
12803496
98%
2-Amino-4-bromopyrimidine
White
MFCD09999168
111375
Slightly soluble in water.
C1=CN=C(N=C1Br)N
174.001
174
Powder
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only