Learn More
2-Amino-4-chloro-5,6-dimethylpyrimidine, 97%
CAS: 14394-61-7 | C6H8ClN3 | 157.601 g/mol
$247.14 - $822.63
Chemical Identifiers
| CAS | 14394-61-7 |
|---|---|
| Molecular Formula | C6H8ClN3 |
| Molecular Weight (g/mol) | 157.601 |
| MDL Number | MFCD00462111 |
| InChI Key | RRMVDLLCVGZDMH-UHFFFAOYSA-N |
| PubChem CID | 248655 |
| IUPAC Name | 4-chloro-5,6-dimethylpyrimidin-2-amine |
| SMILES | CC1=C(N=C(N=C1Cl)N)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5455203
|
Thermo Scientific Chemicals
H5455203 |
1 g |
Each for $247.14
|
|
|||||
|
AAH5455206
|
Thermo Scientific Chemicals
H5455206 |
5 g |
Each for $822.63
|
|
|||||
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Soluble in Chloroform and Ethanol
Notes
Store at -20°C. Keep away from light and heat.
Chemical Identifiers
| 14394-61-7 | |
| 157.601 | |
| RRMVDLLCVGZDMH-UHFFFAOYSA-N | |
| 4-chloro-5,6-dimethylpyrimidin-2-amine |
| C6H8ClN3 | |
| MFCD00462111 | |
| 248655 | |
| CC1=C(N=C(N=C1Cl)N)C |
Specifications
| 14394-61-7 | |
| MFCD00462111 | |
| 4-chloro-5,6-dimethyl-pyrimidin-2-ylamine, 2-amino-4-chloro-5,6-dimethylpyrimidine, 2-pyrimidinamine, 4-chloro-5,6-dimethyl, 2-amino-4-chloro-5,6-dimethyl-pyrimidine, 6-chloro-4,5-dimethylpyrimidine-2-ylamine, 2-pyrimidinamine,4-chloro-5,6-dimethyl | |
| RRMVDLLCVGZDMH-UHFFFAOYSA-N | |
| 4-chloro-5,6-dimethylpyrimidin-2-amine | |
| 248655 | |
| 97% |
| C6H8ClN3 | |
| 1 g | |
| Soluble in Chloroform and Ethanol | |
| CC1=C(N=C(N=C1Cl)N)C | |
| 157.601 | |
| 157.6 | |
| 2-Amino-4-chloro-5,6-dimethylpyrimidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only