Learn More
2-Amino-4-cyanopyridine, 97%
CAS: 42182-27-4 | C6H5N3 | 119.127 g/mol
$89.32 - $1076.84
Chemical Identifiers
| CAS | 42182-27-4 |
|---|---|
| Molecular Formula | C6H5N3 |
| Molecular Weight (g/mol) | 119.127 |
| MDL Number | MFCD03791310 |
| InChI Key | GEEAYLFEIFJFGP-UHFFFAOYSA-N |
| Synonym | 2-amino-4-cyanopyridine, 2-aminoisonicotinonitrile, 2-amino-isonicotinonitrile, pubchem7664, 4-cyanopyridin-2-amine, 4-cyano-2-aminopyridine, acmc-1art6, 2-amino-4-cyano-pyridine, ksc490s3f, 4-cyano-2-pyridinamine |
| PubChem CID | 7015524 |
| IUPAC Name | 2-aminopyridine-4-carbonitrile |
| SMILES | C1=CN=C(C=C1C#N)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1984103
|
Thermo Scientific Chemicals
L1984103 |
1 g |
Each for $89.32
|
|
|||||
|
AAL1984106
|
Thermo Scientific Chemicals
L1984106 |
5 g |
Each for $280.68
|
|
|||||
|
AAL1984114
|
Thermo Scientific Chemicals
L1984114 |
25 g |
Each for $1,076.84
|
|
|||||
Description
2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
| 42182-27-4 | |
| 119.127 | |
| GEEAYLFEIFJFGP-UHFFFAOYSA-N | |
| 7015524 | |
| C1=CN=C(C=C1C#N)N |
| C6H5N3 | |
| MFCD03791310 | |
| 2-amino-4-cyanopyridine, 2-aminoisonicotinonitrile, 2-amino-isonicotinonitrile, pubchem7664, 4-cyanopyridin-2-amine, 4-cyano-2-aminopyridine, acmc-1art6, 2-amino-4-cyano-pyridine, ksc490s3f, 4-cyano-2-pyridinamine | |
| 2-aminopyridine-4-carbonitrile |
Specifications
| 42182-27-4 | |
| C6H5N3 | |
| 1 g | |
| 386393 | |
| Slightly soluble in water. | |
| C1=CN=C(C=C1C#N)N | |
| 119.127 | |
| 119.13 | |
| 2-Amino-4-cyanopyridine |
| 148°C to 152°C | |
| MFCD03791310 | |
| UN3439 | |
| 2-amino-4-cyanopyridine, 2-aminoisonicotinonitrile, 2-amino-isonicotinonitrile, pubchem7664, 4-cyanopyridin-2-amine, 4-cyano-2-aminopyridine, acmc-1art6, 2-amino-4-cyano-pyridine, ksc490s3f, 4-cyano-2-pyridinamine | |
| GEEAYLFEIFJFGP-UHFFFAOYSA-N | |
| 2-aminopyridine-4-carbonitrile | |
| 7015524 | |
| 97% |
Safety and Handling
GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P361-P363-P501c
H301+H311-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only