missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-4'-methoxyacetophenone hydrochloride, 95%, Thermo Scientific™
$96.71 - $96.71
Chemical Identifiers
CAS | 3883-94-1 |
---|---|
Molecular Formula | C9H12ClNO2 |
Molecular Weight (g/mol) | 201.65 |
MDL Number | MFCD00193078 |
InChI Key | FZVYWBMMOSHMRS-UHFFFAOYSA-N |
Synonym | 2-amino-1-4-methoxyphenyl ethanone hydrochloride, 2-amino-4'-methoxyacetophenone hydrochloride, 2-amino-1-4-methoxyphenyl ethan-1-one hydrochloride, 4-methoxyphenacylamine hydrochloride, ethanone, 2-amino-1-4-methoxyphenyl-, hydrochloride, 2-amino-1-4-methoxyphenyl ethan-1-one, chloride, pubchem22350, acmc-1ad40, 4-aminoacetyl anisole hydrochloride, 2-amino-4-methoxyacetophenone hydrochloride |
PubChem CID | 12487188 |
IUPAC Name | 2-amino-1-(4-methoxyphenyl)ethanone;hydrochloride |
SMILES | COC1=CC=C(C=C1)C(=O)CN.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6696603
|
Thermo Scientific Chemicals
H6696603 |
1 g |
Each for $96.71
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3883-94-1 | |
201.65 | |
FZVYWBMMOSHMRS-UHFFFAOYSA-N | |
12487188 | |
COC1=CC=C(C=C1)C(=O)CN.Cl |
C9H12ClNO2 | |
MFCD00193078 | |
2-amino-1-4-methoxyphenyl ethanone hydrochloride, 2-amino-4'-methoxyacetophenone hydrochloride, 2-amino-1-4-methoxyphenyl ethan-1-one hydrochloride, 4-methoxyphenacylamine hydrochloride, ethanone, 2-amino-1-4-methoxyphenyl-, hydrochloride, 2-amino-1-4-methoxyphenyl ethan-1-one, chloride, pubchem22350, acmc-1ad40, 4-aminoacetyl anisole hydrochloride, 2-amino-4-methoxyacetophenone hydrochloride | |
2-amino-1-(4-methoxyphenyl)ethanone;hydrochloride |
Specifications
3883-94-1 | |
White | |
MFCD00193078 | |
3630310 | |
FZVYWBMMOSHMRS-UHFFFAOYSA-N | |
2-amino-1-(4-methoxyphenyl)ethanone;hydrochloride | |
12487188 | |
95% | |
2-Amino-4'-methoxyacetophenone hydrochloride |
190°C to 193°C | |
C9H12ClNO2 | |
1 g | |
2-amino-1-4-methoxyphenyl ethanone hydrochloride, 2-amino-4'-methoxyacetophenone hydrochloride, 2-amino-1-4-methoxyphenyl ethan-1-one hydrochloride, 4-methoxyphenacylamine hydrochloride, ethanone, 2-amino-1-4-methoxyphenyl-, hydrochloride, 2-amino-1-4-methoxyphenyl ethan-1-one, chloride, pubchem22350, acmc-1ad40, 4-aminoacetyl anisole hydrochloride, 2-amino-4-methoxyacetophenone hydrochloride | |
COC1=CC=C(C=C1)C(=O)CN.Cl | |
201.65 | |
201.65 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures