missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-methoxybenzothiazole 98.0+%, TCI America™
$149.13 - $538.71
Chemical Identifiers
| CAS | 5464-79-9 |
|---|---|
| Molecular Formula | C8H8N2OS |
| Molecular Weight (g/mol) | 180.225 |
| MDL Number | MFCD00005792 |
| InChI Key | YEBCRAVYUWNFQT-UHFFFAOYSA-N |
| Synonym | 2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci |
| PubChem CID | 21622 |
| IUPAC Name | 4-methoxy-1,3-benzothiazol-2-amine |
| SMILES | COC1=C2C(=CC=C1)SC(=N2)N |
Chemical Identifiers
| 5464-79-9 | |
| 180.225 | |
| YEBCRAVYUWNFQT-UHFFFAOYSA-N | |
| 21622 | |
| COC1=C2C(=CC=C1)SC(=N2)N |
| C8H8N2OS | |
| MFCD00005792 | |
| 2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci | |
| 4-methoxy-1,3-benzothiazol-2-amine |
Specifications
| 5464-79-9 | |
| White-Yellow | |
| MFCD00005792 | |
| 2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci | |
| COC1=C2C(=CC=C1)SC(=N2)N | |
| 180.225 | |
| 180.23 | |
| Crystalline Powder |
| 156°C | |
| C8H8N2OS | |
| 1 g | |
| YEBCRAVYUWNFQT-UHFFFAOYSA-N | |
| 4-methoxy-1,3-benzothiazol-2-amine | |
| 21622 | |
| ≥98.0% (HPLC) | |
| 2-Amino-4-methoxybenzothiazole |
Safety and Handling
RTECSNumber : DL2000000
TSCA : No