missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-nitrophenol Sodium Salt 80.0+%, TCI America™

Supplier:  TCI America A0380500G

 View more versions of this product

Catalog No. A0380500G


Description

Description

Specifications

Specifications

2-Amino-4-nitrophenol Sodium Salt
C6H5N2NaO3
500 g
QROXVHUAKDVYQE-UHFFFAOYSA-M
sodium;2-amino-4-nitrophenolate
5485279
≥80.0% (T)
61702-43-0
MFCD00067428
2-Hydroxy-5-nitroaniline Sodium Salt
C1=CC(=C(C=C1[N+](=O)[O-])N)[O-].[Na+]
176.107
176.11
Crystalline Powder

Chemical Identifiers

61702-43-0
176.107
QROXVHUAKDVYQE-UHFFFAOYSA-M
5485279
C1=CC(=C(C=C1[N+](=O)[O-])N)[O-].[Na+]
C6H5N2NaO3
MFCD00067428
2-Hydroxy-5-nitroaniline Sodium Salt
sodium;2-amino-4-nitrophenolate
Safety and Handling

Safety and Handling

RTECSNumber : SJ6305000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions