missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-bromopyrimidine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 7752-82-1
Molecular Formula C4H4BrN3
Molecular Weight (g/mol) 174
MDL Number MFCD00012341
InChI Key UHRHPPKWXSNZLR-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyrimidine, 5-bromopyrimidin-2-ylamine, 2-pyrimidinamine, 5-bromo, 5-bromo-2-aminopyrimidine, 5-bromo-2-pyrimidinamine, pyrimidine, 2-amino-5-bromo, 5-bromopyrimidine-2-amine, 2-amino-5-bromo-pyrimidine, 5-bromopyrimidine-2-ylamine, 5-bromo-pyrimidin-2-ylamine
PubChem CID 231310
IUPAC Name 5-bromopyrimidin-2-amine
SMILES C1=C(C=NC(=N1)N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC368580010
SDS View Documents
Thermo Scientific Chemicals
368580010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC368580050
SDS View Documents
Thermo Scientific Chemicals
368580050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7752-82-1
174
UHRHPPKWXSNZLR-UHFFFAOYSA-N
231310
C1=C(C=NC(=N1)N)Br
C4H4BrN3
MFCD00012341
2-amino-5-bromopyrimidine, 5-bromopyrimidin-2-ylamine, 2-pyrimidinamine, 5-bromo, 5-bromo-2-aminopyrimidine, 5-bromo-2-pyrimidinamine, pyrimidine, 2-amino-5-bromo, 5-bromopyrimidine-2-amine, 2-amino-5-bromo-pyrimidine, 5-bromopyrimidine-2-ylamine, 5-bromo-pyrimidin-2-ylamine
5-bromopyrimidin-2-amine

Specifications

7752-82-1
100.0
Authentic
Glass bottle
MFCD00012341
25, IV, 2125
Solubility in water: insoluble
C1=C(C=NC(=N1)N)Br
174
174
2-Amino-5-bromopyrimidine, 97%
96.0
>300°C
96% min. (HPLC)
C4H4BrN3
1g
2-amino-5-bromopyrimidine, 5-bromopyrimidin-2-ylamine, 2-pyrimidinamine, 5-bromo, 5-bromo-2-aminopyrimidine, 5-bromo-2-pyrimidinamine, pyrimidine, 2-amino-5-bromo, 5-bromopyrimidine-2-amine, 2-amino-5-bromo-pyrimidine, 5-bromopyrimidine-2-ylamine, 5-bromo-pyrimidin-2-ylamine
UHRHPPKWXSNZLR-UHFFFAOYSA-N
5-bromopyrimidin-2-amine
231310
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only