Learn More
2-Amino-5-chlorobenzophenone, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1572022
Description
It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.
Solubility
Solubility in methanol gives very faint turbidity. Soluble in DMSO.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
2-Amino-5-chlorobenzophenone | |
96°C to 99°C | |
211°C (412°F) | |
MFCD00007839 | |
475640 | |
ZUWXHHBROGLWNH-UHFFFAOYSA-N | |
(2-amino-5-chlorophenyl)-phenylmethanone | |
12870 | |
≥98% |
719-59-5 | |
1.33 | |
C13H10ClNO | |
100 g | |
2-amino-5-chlorobenzophenone, 2-amino-5-chlorophenyl phenyl methanone, 2-benzoyl-4-chloroaniline, methanone, 2-amino-5-chlorophenyl phenyl, 2-amino-5-chlorbenzophenone, 5-chloro-2-aminobenzophenone, 2-amino-5-chlorobenzylphenone, benzophenone, 2-amino-5-chloro, unii-fr80014zbt, 2-amino-5-chlorophenyl phenylmethanone | |
NC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1 | |
231.68 | |
231.68 |
Chemical Identifiers
719-59-5 | |
231.68 | |
ZUWXHHBROGLWNH-UHFFFAOYSA-N | |
12870 | |
NC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1 |
C13H10ClNO | |
MFCD00007839 | |
2-amino-5-chlorobenzophenone, 2-amino-5-chlorophenyl phenyl methanone, 2-benzoyl-4-chloroaniline, methanone, 2-amino-5-chlorophenyl phenyl, 2-amino-5-chlorbenzophenone, 5-chloro-2-aminobenzophenone, 2-amino-5-chlorobenzylphenone, benzophenone, 2-amino-5-chloro, unii-fr80014zbt, 2-amino-5-chlorophenyl phenylmethanone | |
(2-amino-5-chlorophenyl)-phenylmethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-949-7
RTECSNumber : PC4933500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only