missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-fluorobenzamide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6687303

 View more versions of this product

Catalog No. AAH6687303


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-5-fluorobenzamide
143°C to 148°C
C7H7FN2O
1 g
2-amino-5-fluoro-benzamide, benzamide, 2-amino-5-fluoro, acmc-209nco, 5-fluoroanthranilamide, benzamide,2-amino-5-fluoro
C1=CC(=C(C=C1F)C(=O)N)N
154.144
154.14
Crystalline
63069-49-8
White
MFCD03428522
2718500
RHJMYIPLYKQZJM-UHFFFAOYSA-N
2-amino-5-fluorobenzamide
10241003
97%

Chemical Identifiers

63069-49-8
154.144
RHJMYIPLYKQZJM-UHFFFAOYSA-N
2-amino-5-fluorobenzamide
C7H7FN2O
MFCD03428522
10241003
C1=CC(=C(C=C1F)C(=O)N)N
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only