missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-5-fluorophenol 97.0+%, TCI America™
$72.67 - $211.08
Chemical Identifiers
CAS | 53981-24-1 |
---|---|
Molecular Formula | C6H6FNO |
Molecular Weight (g/mol) | 127.118 |
MDL Number | MFCD00671759 |
InChI Key | IIDUNAVOCYMUFB-UHFFFAOYSA-N |
Synonym | 4-fluoro-2-hydroxyaniline, phenol, 2-amino-5-fluoro, 2-amino-5-fluoro-phenol, 5-fluoro-2-aminophenol, 2-hydroxy-4-fluoroaniline, pubchem2828, acmc-1avob, 2-amino-5-flourophenol, 2-amino-5-fluor-phenol, 2-amino-5-fluoro phenol |
PubChem CID | 185763 |
IUPAC Name | 2-amino-5-fluorophenol |
SMILES | C1=CC(=C(C=C1F)O)N |
Chemical Identifiers
53981-24-1 | |
127.118 | |
IIDUNAVOCYMUFB-UHFFFAOYSA-N | |
185763 | |
C1=CC(=C(C=C1F)O)N |
C6H6FNO | |
MFCD00671759 | |
4-fluoro-2-hydroxyaniline, phenol, 2-amino-5-fluoro, 2-amino-5-fluoro-phenol, 5-fluoro-2-aminophenol, 2-hydroxy-4-fluoroaniline, pubchem2828, acmc-1avob, 2-amino-5-flourophenol, 2-amino-5-fluor-phenol, 2-amino-5-fluoro phenol | |
2-amino-5-fluorophenol |
Specifications
53981-24-1 | |
MFCD00671759 | |
4-fluoro-2-hydroxyaniline, phenol, 2-amino-5-fluoro, 2-amino-5-fluoro-phenol, 5-fluoro-2-aminophenol, 2-hydroxy-4-fluoroaniline, pubchem2828, acmc-1avob, 2-amino-5-flourophenol, 2-amino-5-fluor-phenol, 2-amino-5-fluoro phenol | |
C1=CC(=C(C=C1F)O)N | |
127.118 | |
127.12 | |
Crystalline Powder |
C6H6FNO | |
1 g | |
IIDUNAVOCYMUFB-UHFFFAOYSA-N | |
2-amino-5-fluorophenol | |
185763 | |
≥97.0% (GC) | |
2-Amino-5-fluorophenol |
Safety and Handling
TSCA : No