missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-iodopyrimidine, 97%, Thermo Scientific Chemicals

$65.00 - $704.90

Chemical Identifiers

CAS 1445-39-2
Molecular Formula C4H4IN3
Molecular Weight (g/mol) 221.001
MDL Number MFCD01075666
InChI Key HAFKCGZQRIIADX-UHFFFAOYSA-N
Synonym 2-amino-5-iodopyrimidine, 2-amino-5-iodo-pyrimidine, 5-iodo-pyrimidin-2-ylamine, 5-iodopyrimidine-2-ylamine, zlchem 484, 5-iodo-2-pyrimidinamine, 2-pyrimidinamine, 5-iodo, acmc-1c050
PubChem CID 241102
IUPAC Name 5-iodopyrimidin-2-amine
SMILES C1=C(C=NC(=N1)N)I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2270503
View Documents
Thermo Scientific Chemicals
B2270503
1 g
Each for $65.00
 
AAB2270506
View Documents
Thermo Scientific Chemicals
B2270506
5 g
Each for $204.53
 
AAB2270514
View Documents
Thermo Scientific Chemicals
B2270514
25 g
Each for $704.90
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1445-39-2
221.001
HAFKCGZQRIIADX-UHFFFAOYSA-N
241102
C1=C(C=NC(=N1)N)I
C4H4IN3
MFCD01075666
2-amino-5-iodopyrimidine, 2-amino-5-iodo-pyrimidine, 5-iodo-pyrimidin-2-ylamine, 5-iodopyrimidine-2-ylamine, zlchem 484, 5-iodo-2-pyrimidinamine, 2-pyrimidinamine, 5-iodo, acmc-1c050
5-iodopyrimidin-2-amine

Specifications

1445-39-2
C4H4IN3
1 g
2-amino-5-iodopyrimidine, 2-amino-5-iodo-pyrimidine, 5-iodo-pyrimidin-2-ylamine, 5-iodopyrimidine-2-ylamine, zlchem 484, 5-iodo-2-pyrimidinamine, 2-pyrimidinamine, 5-iodo, acmc-1c050
C1=C(C=NC(=N1)N)I
221.001
221
2-Amino-5-iodopyrimidine
217°C to 220°C
MFCD01075666
Light sensitive
HAFKCGZQRIIADX-UHFFFAOYSA-N
5-iodopyrimidin-2-amine
241102
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only